missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-172846
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
612-23-7
C7H6ClNO2
171.58
MFCD00007185
BXCBUWKTXLWPSB-UHFFFAOYSA-N
2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
11921
1-(chloromethyl)-2-nitrobenzene
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]
This item is not returnable. View return policy
612-23-7
C7H6ClNO2
171.58
MFCD00007185
BXCBUWKTXLWPSB-UHFFFAOYSA-N
2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
11921
1-(chloromethyl)-2-nitrobenzene
C1=CC=C(C(=C1)CCl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 44°C to 49°C
Color Tan to Yellow
Boiling Point 127°C to 133°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2Cl
Quantity 10 g
Beilstein 05, 327
Solubility Information Solubility in water: insoluble in water
Formula Weight 171.58
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-Nitrobenzyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'