missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl acetate, 99%, Thermo Scientific Chemicals

Catalog Number p-7024183
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
1523-11-1
C12H10O2
186.21
MFCD00004060
RJNPPEUAJCEUPV-UHFFFAOYSA-N
2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
73709
naphthalen-2-yl acetate
CC(=O)OC1=CC=C2C=CC=CC2=C1
This item is not returnable. View return policy
1523-11-1
C12H10O2
186.21
MFCD00004060
RJNPPEUAJCEUPV-UHFFFAOYSA-N
2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
73709
naphthalen-2-yl acetate
CC(=O)OC1=CC=C2C=CC=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 70°C
Quantity 10 g
Beilstein 1868291
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Naphthyl acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'