Learn More
2-Morpholinophenol, 97%, Thermo Scientific™
Catalog Number p-2698686
Click to view available options
Quantity:
1 g
10 g
41536-44-1
C10H13NO2
179.219
MFCD00051674
VMPYFWTYGZZUMY-UHFFFAOYSA-N
2795394
2-morpholin-4-ylphenol
C1COCCN1C2=CC=CC=C2O
41536-44-1
C10H13NO2
179.219
MFCD00051674
VMPYFWTYGZZUMY-UHFFFAOYSA-N
2795394
2-morpholin-4-ylphenol
C1COCCN1C2=CC=CC=C2O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-morpholinophenol, 2-morpholin-4-yl phenol, 2-4-morpholino phenol, phenol, 2-4-morpholinyl, 2-4-morpholinyl phenol, 2-morpholin-4-yl-phenol, 2-4-morpholinyl-phenol, phenol,2-4-morpholinyl
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 2-Morpholinophenol
compliance-icons
Product Identifier
  • 2-Morpholinophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.