Learn More
2-Morpholinoaniline, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifica
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 178.23 |
Percent Purity | 97% |
Chemical Name or Material | 2-Morpholinoaniline |