missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)phenyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number p-7045630
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51333-75-6
C8H7NS2
181.271
MFCD00041061
SIODOISHDGRELW-UHFFFAOYSA-N
2-methylthio phenyl isothiocyanate, 1-isothiocyanato-2-methylsulfanyl benzene, acmc-20aomj, 2-methylthiobenzenisothiocyanate, 2-methylthio-phenyl-isothiocyanate, 1-isothiocyanato-2-methylthio benzene, 2-methylthio phenylisothiocyanate, 1-isothiocyanato-2-methylsulfanyl-benzene, benzene,1-isothiocyanato-2-methylthio
142840
1-isothiocyanato-2-methylsulfanylbenzene
CSC1=CC=CC=C1N=C=S
This item is not returnable. View return policy
51333-75-6
C8H7NS2
181.271
MFCD00041061
SIODOISHDGRELW-UHFFFAOYSA-N
2-methylthio phenyl isothiocyanate, 1-isothiocyanato-2-methylsulfanyl benzene, acmc-20aomj, 2-methylthiobenzenisothiocyanate, 2-methylthio-phenyl-isothiocyanate, 1-isothiocyanato-2-methylthio benzene, 2-methylthio phenylisothiocyanate, 1-isothiocyanato-2-methylsulfanyl-benzene, benzene,1-isothiocyanato-2-methylthio
142840
1-isothiocyanato-2-methylsulfanylbenzene
CSC1=CC=CC=C1N=C=S

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.227
Boiling Point 149°C to 150°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.695
Quantity 1 g
UN Number UN2922
Beilstein 3243502
Sensitivity Moisture sensitive
Formula Weight 181.28
Percent Purity 97%
Chemical Name or Material 2-(Methylthio)phenyl isothiocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'