missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine-4-boronic acid pinacol ester, 95%, may contain up to 3% water, Thermo Scientific™

Catalog Number p-7029928
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
660867-80-1
C12H18BNO2
219.09
MFCD08061590
MBTULFIFECUURA-UHFFFAOYSA-N
2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester
16414274
2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
660867-80-1
C12H18BNO2
219.09
MFCD08061590
MBTULFIFECUURA-UHFFFAOYSA-N
2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester
16414274
2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 66°C to 71°C
Quantity 250 mg
Formula Weight 219.09
Percent Purity 95%
Assay may contain up to 3% water
Chemical Name or Material 2-Methylpyridine-4-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'