missing translation for 'onlineSavingsMsg'
Learn More

2-Methylmalonamide, 95%, Thermo Scientific Chemicals

Catalog Number p-7045039
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
1113-63-9
C4H8N2O2
116.12
MFCD00043625
CGRKYGKHZOCPSZ-UHFFFAOYSA-N
2-methylmalonamide, 2-methylmalonodiamide, methylmalonamide, methyl malonamide, 2-methyl malonamide, acmc-20anpx, propanediamide, 2-methyl
227804
2-methylpropanediamide
CC(C(N)=O)C(N)=O
This item is not returnable. View return policy
1113-63-9
C4H8N2O2
116.12
MFCD00043625
CGRKYGKHZOCPSZ-UHFFFAOYSA-N
2-methylmalonamide, 2-methylmalonodiamide, methylmalonamide, methyl malonamide, 2-methyl malonamide, acmc-20anpx, propanediamide, 2-methyl
227804
2-methylpropanediamide
CC(C(N)=O)C(N)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 208°C to 210°C
Quantity 2 g
Beilstein 1756100
Formula Weight 116.12
Percent Purity 95%
Chemical Name or Material 2-Methylmalonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'