missing translation for 'onlineSavingsMsg'
Learn More

2-Methylfuran, 98+%, stab., Thermo Scientific Chemicals

Catalog Number p-7027154
Click to view available options
Quantity:
250 mL
1000 mL
This item is not returnable. View return policy
534-22-5
C5H6O
82.102
MFCD00003248
VQKFNUFAXTZWDK-UHFFFAOYSA-N
methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
10797
2-methylfuran
CC1=CC=CO1
This item is not returnable. View return policy
534-22-5
C5H6O
82.102
MFCD00003248
VQKFNUFAXTZWDK-UHFFFAOYSA-N
methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
10797
2-methylfuran
CC1=CC=CO1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 89°C
Density 0.910
Boiling Point 63°C to 65°C
Flash Point -30°C (-22°F)
Assay Percent Range 98+%
Refractive Index 1.433
Quantity 250 mL
UN Number UN2301
Beilstein 103733
Formula Weight 82.1
Percent Purity ≥98%
Chemical Name or Material 2-Methylfuran, stab. with 150-400ppm BHT
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'