missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(4-morpholinyl)benzenamine, 97%, Thermo Scientific Chemicals

Catalog Number p-4228782
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
581-00-0
C11H16N2O
192.26
MFCD10686817
ZGJUJDQANIYVAL-UHFFFAOYSA-N
2-methyl-4-morpholin-4-yl aniline, 2-methyl-4-morpholinoaniline, 2-methyl-4-4-morpholinyl aniline, 4-p-amino-m-tolyl morpholine, 4-4-morpholinyl-2-methylaniline, 2-methyl4-4-morpholinyl benzenamine, n-4-amino-3-methylphenyl morpholine, 2-methyl-4-4-morpholinyl benzenamine, 2-methyl-4-morpholin-4-yl-phenylamine, benzenamine, 2-methyl-4-4-morpholinyl
21955000
2-methyl-4-morpholin-4-ylaniline
CC1=C(C=CC(=C1)N2CCOCC2)N
This item is not returnable. View return policy
581-00-0
C11H16N2O
192.26
MFCD10686817
ZGJUJDQANIYVAL-UHFFFAOYSA-N
2-methyl-4-morpholin-4-yl aniline, 2-methyl-4-morpholinoaniline, 2-methyl-4-4-morpholinyl aniline, 4-p-amino-m-tolyl morpholine, 4-4-morpholinyl-2-methylaniline, 2-methyl4-4-morpholinyl benzenamine, n-4-amino-3-methylphenyl morpholine, 2-methyl-4-4-morpholinyl benzenamine, 2-methyl-4-morpholin-4-yl-phenylamine, benzenamine, 2-methyl-4-4-morpholinyl
21955000
2-methyl-4-morpholin-4-ylaniline
CC1=C(C=CC(=C1)N2CCOCC2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 92°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 192.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-(4-morpholinyl)benzenamine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-4-(4-morpholinyl)benzenamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'