missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-phenylpropanenitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-4228820
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1195-98-8
C10H11N
145.21
PGQTYXFMSZUGOW-UHFFFAOYSA-N
2-methyl-2-phenyl-propionitrile, phenylisobutyronitrile, 2-phenylisobutyronitrile, 2-methyl-2-phenylpropionitrile, 2-methyl-2-phenyl-propanenitrile, alpha,alpha-dimethylbenzyl cyanide, benzeneacetonitrile,a,a-dimethyl, alpha,alpha-dimethylphenylacetonitrile, benzeneacetonitrile,, a,, a-dimethyl, benzeneacetonitrile, alpha,alpha-dimethyl
2797591
2-methyl-2-phenylpropanenitrile
CC(C)(C#N)C1=CC=CC=C1
This item is not returnable. View return policy
1195-98-8
C10H11N
145.21
PGQTYXFMSZUGOW-UHFFFAOYSA-N
2-methyl-2-phenyl-propionitrile, phenylisobutyronitrile, 2-phenylisobutyronitrile, 2-methyl-2-phenylpropionitrile, 2-methyl-2-phenyl-propanenitrile, alpha,alpha-dimethylbenzyl cyanide, benzeneacetonitrile,a,a-dimethyl, alpha,alpha-dimethylphenylacetonitrile, benzeneacetonitrile,, a,, a-dimethyl, benzeneacetonitrile, alpha,alpha-dimethyl
2797591
2-methyl-2-phenylpropanenitrile
CC(C)(C#N)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 54°C to 56°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 145.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-phenylpropanenitrile
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2-phenylpropanenitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'