Learn More
2-Methyl-1H-imidazole-4-carbothioamide, 97%, Thermo Scientific™
Catalog Number p-2698591
Click to view available options
Quantity:
250 mg
1 g
5 g
129486-91-5
C5H7N3S
141.192
MFCD03659724
MNYPQSNAWPZXRV-UHFFFAOYSA-N
2-methyl-1h-imidazole-4-carbothioamide, 1h-imidazole-5-carbothioamide,2-methyl, 1h-imidazole-5-carbothioamide, 2-methyl, acmc-1c27g, amino 2-methylimidazol-4-yl methane-1-thione, 2-methyl-3h-imidazole-4-carbothioamide
2796714
2-methyl-1H-imidazole-5-carbothioamide
CC1=NC=C(N1)C(=S)N
129486-91-5
C5H7N3S
141.192
MFCD03659724
MNYPQSNAWPZXRV-UHFFFAOYSA-N
2-methyl-1h-imidazole-4-carbothioamide, 1h-imidazole-5-carbothioamide,2-methyl, 1h-imidazole-5-carbothioamide, 2-methyl, acmc-1c27g, amino 2-methylimidazol-4-yl methane-1-thione, 2-methyl-3h-imidazole-4-carbothioamide
2796714
2-methyl-1H-imidazole-5-carbothioamide
CC1=NC=C(N1)C(=S)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 141.2
Percent Purity 97%
Chemical Name or Material 2-Methyl-1H-imidazole-4-carbothioamide