Learn More
2-Methoxytetrahydropyran, 98%, Thermo Scientific Chemicals
Catalog Number p-4812820
Click to view available options
Quantity:
5 g
25 g
6581-66-4
C6H12O2
116.16
XTDKZSUYCXHXJM-UHFFFAOYSA-N
2-methoxytetrahydropyran, tetrahydro-2-methoxy-2h-pyran, pyran, 2-methoxy-tetrahydro, 2h-pyran,tetrahydro-2-methoxy, 2h-pyran, tetrahydro-2-methoxy, 2-methoxytetrahydro-2h-pyran, methoxytetrahydropyran, 2-methoxyoxan-3-yl, 5-17-03-00078 beilstein handbook reference
23057
2-methoxyoxane
COC1CCCCO1
6581-66-4
C6H12O2
116.16
XTDKZSUYCXHXJM-UHFFFAOYSA-N
2-methoxytetrahydropyran, tetrahydro-2-methoxy-2h-pyran, pyran, 2-methoxy-tetrahydro, 2h-pyran,tetrahydro-2-methoxy, 2h-pyran, tetrahydro-2-methoxy, 2-methoxytetrahydro-2h-pyran, methoxytetrahydropyran, 2-methoxyoxan-3-yl, 5-17-03-00078 beilstein handbook reference
23057
2-methoxyoxane
COC1CCCCO1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.9630g/mL
Boiling Point 128°C to 129°C
Flash Point 25°C
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 0.963
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 2-Methoxytetrahydropyran
compliance-icons
Product Identifier
  • 2-Methoxytetrahydropyran, 98%
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.