Learn More
2-Methoxypyridine-5-boronic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
Specifications
Melting Point | 132°C to 133°C |
Quantity | 250 mg |
Beilstein | 9119950 |
Solubility Information | Soluble in water. |
Formula Weight | 152.95 |
Percent Purity | 95% |
Chemical Name or Material | 2-Methoxypyridine-5-boronic acid |