Learn More
Alfa Aesar™ 2-Methoxy-5-(trifluoromethyl)benzonitrile, 97%
Brand: Alfa Aesar™ H26621.14
Additional Details : CAS Number : 34636-92-5 Weight : 0.02500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
34636-92-5 | |
201.148 | |
KAJZODLWVHKBSI-UHFFFAOYSA-N | |
17750808 | |
COC1=C(C=C(C=C1)C(F)(F)F)C#N |
C9H6F3NO | |
MFCD06409082 | |
2-methoxy-5-trifluoromethyl benzonitrile, acmc-1agd1, 2-cyano-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl-benzonitrile, benzonitrile,2-methoxy-5-trifluoromethyl, benzonitrile, 2-methoxy-5-trifluoromethyl, 2-methoxy-5-trifluoromethyl benzenecarbonitrile | |
2-methoxy-5-(trifluoromethyl)benzonitrile |
Specifications
2-Methoxy-5-(trifluoromethyl)benzonitrile | |
25g | |
C9H6F3NO | |
UN3439 | |
KAJZODLWVHKBSI-UHFFFAOYSA-N | |
2-methoxy-5-(trifluoromethyl)benzonitrile | |
17750808 | |
97% |
30°C to 34°C | |
34636-92-5 | |
MFCD06409082 | |
2-methoxy-5-trifluoromethyl benzonitrile, acmc-1agd1, 2-cyano-4-trifluoromethyl anisole, 2-methoxy-5-trifluoromethyl-benzonitrile, benzonitrile,2-methoxy-5-trifluoromethyl, benzonitrile, 2-methoxy-5-trifluoromethyl, 2-methoxy-5-trifluoromethyl benzenecarbonitrile | |
COC1=C(C=C(C=C1)C(F)(F)F)C#N | |
201.148 | |
201.15 |