missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptoethane sulfonic acid sodium salt, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
Description
2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.
Solubility
Soluble in water. Also soluble in methanol, DMSO (33 mg/ml) and ethanol (<1mg/mL).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.
Solubility
Soluble in water. Also soluble in methanol, DMSO (33 mg/ml) and ethanol (<1mg/mL).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Color | White |
Linear Formula | HSCH2CH2SO3Na |
Quantity | 10 g |
Beilstein | 3657828 |
Merck Index | 14,5909 |
Solubility Information | Soluble in water. Also soluble in methanol,DMSO (33mg/ml) and ethanol (<1mg/ml). |
Formula Weight | 164.18 |
Percent Purity | 96% |
Physical Form | Powder |
Chemical Name or Material | 2-Mercaptoethane sulfonic acid sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'