missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7021667
182.40 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
100 g
500 g
2500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
149-30-4
C7H5NS2
167.244
MFCD00005781
YXIWHUQXZSMYRE-UHFFFAOYSA-N
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
697993
CHEBI:34292
3H-1,3-benzothiazole-2-thione
C1=CC=C2C(=C1)NC(=S)S2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
149-30-4
C7H5NS2
167.244
MFCD00005781
YXIWHUQXZSMYRE-UHFFFAOYSA-N
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
697993
CHEBI:34292
3H-1,3-benzothiazole-2-thione
C1=CC=C2C(=C1)NC(=S)S2

2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.

Solubility
Soluble in ethanol, diethyl ether, benzene, carbon tetrachloride, alkali and alkaline solutions. Slightly soluble in acetone, naphtha and glacial acetic acid. Insoluble in water and gasoline.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 178°C to 182°C
Flash Point 243°C (469°F)
Quantity 10 g
UN Number UN3077
Beilstein 119484
Merck Index 14,5868
Solubility Information Soluble in alkali and alkali carbonate solutions
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'