missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole, 95%, Thermo Scientific Chemicals

Catalog Number p-3758284
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
149-30-4
C7H5NS2
167.24
MFCD00005781
YXIWHUQXZSMYRE-UHFFFAOYSA-N
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
697993
CHEBI:34292
3H-1,3-benzothiazole-2-thione
C1=CC=C2C(=C1)NC(=S)S2
This item is not returnable. View return policy
149-30-4
C7H5NS2
167.24
MFCD00005781
YXIWHUQXZSMYRE-UHFFFAOYSA-N
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
697993
CHEBI:34292
3H-1,3-benzothiazole-2-thione
C1=CC=C2C(=C1)NC(=S)S2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 172.0°C to 182.0°C
Color Yellow
Flash Point 243°C
Loss on Drying 0.5% max. (60°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 10 g
Beilstein 27, II, 233
Merck Index 15, 5935
Solubility Information Solubility in water: 0.12g/L (25°C). Other solubilities: soluble in alkalies,soluble in alcohol,acetone,benzene and,chloroform
Formula Weight 167.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'