missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Isopropoxypyridine-3-boronic acid pinacol ester, 96%
Click to view available options
Quantity:
1 g
5 g
Description
Applications
2-Isopropoxypyridine-3-boronic acid pinacol ester is used as an intermediate in the pharmaceutical industry.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
2-Isopropoxypyridine-3-boronic acid pinacol ester is used as an intermediate in the pharmaceutical industry.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Quantity | 1 g |
Solubility Information | Slightly miscible with water. |
Formula Weight | 263.15 |
Percent Purity | 96% |
Chemical Name or Material | 2-Isopropoxypyridine-3-boronic acid pinacol ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'