Learn More
2-Hydroxyquinoline-4-carboxylic acid, 98%, Thermo Scientific™
Catalog Number p-7030509
Click to view available options
Quantity:
1 g
5 g
15733-89-8
C10H7NO3
189.17
MFCD00023942 MFCD00464512
MFSHNFBQNVGXJX-UHFFFAOYSA-N
2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
85076
CHEBI:52045
2-oxo-1H-quinoline-4-carboxylic acid
OC(=O)C1=CC(=O)NC2=CC=CC=C12
15733-89-8
C10H7NO3
189.17
MFCD00023942 MFCD00464512
MFSHNFBQNVGXJX-UHFFFAOYSA-N
2-hydroxyquinoline-4-carboxylic acid, 2-oxo-1,2-dihydroquinoline-4-carboxylic acid, 2-hydroxy-4-quinolincarboxylic acid, 2-hydroxycinchoninic acid, cinchoninic acid, 2-hydroxy, 4-carboxycarbostyril, 2-oxocinchoninic acid, 4-quinolinecarboxylic acid, 1,2-dihydro-2-oxo, 2-hydroxy-quinoline-4-carboxylic acid, 2-hydroxy-4-quinolinecarboxylic acid
85076
CHEBI:52045
2-oxo-1H-quinoline-4-carboxylic acid
OC(=O)C1=CC(=O)NC2=CC=CC=C12
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 189.17
Percent Purity 98%
Chemical Name or Material 2-Hydroxyquinoline-4-carboxylic acid