Learn More
2-Hydroxycarbazole, 97%, Thermo Scientific Chemicals
Catalog Number p-7030212
Click to view available options
Quantity:
5 g
25 g
86-79-3
C12H9NO
183.21
MFCD00004962
GWPGDZPXOZATKL-UHFFFAOYSA-N
2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole
93551
9H-carbazol-2-ol
OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1
86-79-3
C12H9NO
183.21
MFCD00004962
GWPGDZPXOZATKL-UHFFFAOYSA-N
2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole
93551
9H-carbazol-2-ol
OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 262°C to 273°C
Odor Odorless
Quantity 5 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2-Hydroxycarbazole