Learn More
Trajan™ 2-Hole Ferrules
15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems.
Brand: Trajan™ 072674
Additional Details : Weight : 0.01000kg
Description
- Composite of graphite and Vespel
- Mechanically robust, hard material, long lifetime
- Forms a strong grip with the capillary column
Specifications
0.25 in. | |
GC | |
Graphite, Vespel | |
2-hole Ferrule |
1/8 in. Fittings | |
6.4 mm | |
7.4 mm | |
2 x 0.8 mm |