missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7026216
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7304-32-7
C7H4FNO4
185.11
MFCD00134238
ICXSHFWYCHJILC-UHFFFAOYSA-N
2-fluoro-5-nitrobenzoic acid, 2-fluoro-5-nitrobenzoicacid, 2-fluoro-5-nitro-benzoic acid, benzoic acid, 2-fluoro-5-nitro, 2-fluoro-5-nitro-benzoicacid, pubchem1307, acmc-209op6, ksc377e3h, 2-fluoro-5-nitrobenzotc acid, 2-fluoro-5-nitro benzoic acid
280997
2-fluoro-5-nitrobenzoic acid
OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O
This item is not returnable. View return policy
7304-32-7
C7H4FNO4
185.11
MFCD00134238
ICXSHFWYCHJILC-UHFFFAOYSA-N
2-fluoro-5-nitrobenzoic acid, 2-fluoro-5-nitrobenzoicacid, 2-fluoro-5-nitro-benzoic acid, benzoic acid, 2-fluoro-5-nitro, 2-fluoro-5-nitro-benzoicacid, pubchem1307, acmc-209op6, ksc377e3h, 2-fluoro-5-nitrobenzotc acid, 2-fluoro-5-nitro benzoic acid
280997
2-fluoro-5-nitrobenzoic acid
OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O

2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support, synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols, in the synthesis of oxazepines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support, synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols, in the synthesis of oxazepines.

Solubility
Soluble in chloroform, ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 142°C to 144°C
Quantity 1 g
Beilstein 1912835
Solubility Information Soluble in chloroform,ethanol.
Formula Weight 185.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'