missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxybenzenesulfonyl chloride, 96%, Thermo Scientific Chemicals
729.63 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
2-Ethoxybenzenesulfonyl chloride is used as pharmaceutical intermediates.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture, bases.
Specifications
Melting Point | 60°C to 64°C |
Quantity | 1 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Formula Weight | 220.67 |
Percent Purity | 96% |
Chemical Name or Material | 2-Ethoxybenzenesulfonyl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'