missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-7027216
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
97914-59-5
C8H5F2O3
187.12
MFCD00236223
AGDOJFCUKQMLHD-UHFFFAOYSA-M
2-difluoromethoxy benzoic acid, benzoic acid, 2-difluoromethoxy, 2-difluoromethoxy-benzoic acid, rarechem al bo 1907, timtec-bb sbb029907, art-chem-bb b006808, benzoic acid,2-difluoromethoxy, 2-carboxy-alpha,alpha-difluoroanisole, 2-bis fluoranyl methoxy benzoic acid
2063348
2-(difluoromethoxy)benzoic acid
[O-]C(=O)C1=CC=CC=C1OC(F)F
This item is not returnable. View return policy
97914-59-5
C8H5F2O3
187.12
MFCD00236223
AGDOJFCUKQMLHD-UHFFFAOYSA-M
2-difluoromethoxy benzoic acid, benzoic acid, 2-difluoromethoxy, 2-difluoromethoxy-benzoic acid, rarechem al bo 1907, timtec-bb sbb029907, art-chem-bb b006808, benzoic acid,2-difluoromethoxy, 2-carboxy-alpha,alpha-difluoroanisole, 2-bis fluoranyl methoxy benzoic acid
2063348
2-(difluoromethoxy)benzoic acid
[O-]C(=O)C1=CC=CC=C1OC(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 100°C
Quantity 1 g
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'