missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentyloxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7033098
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1073371-90-0
C16H24BNO3
289.18
MFCD07781170
GZCZGFSEHAZEPS-UHFFFAOYSA-N
2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxypyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy pyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester, 2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester
24208792
2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1
This item is not returnable. View return policy
1073371-90-0
C16H24BNO3
289.18
MFCD07781170
GZCZGFSEHAZEPS-UHFFFAOYSA-N
2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxypyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy pyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester, 2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester
24208792
2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 289.19
Percent Purity 96%
Chemical Name or Material 2-Cyclopentyloxypyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'