missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055812
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
95727-86-9
C7H3F3N2
172.11
MFCD01001119
WDSCJULUXJSJOX-UHFFFAOYSA-N
5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
1535372
5-(trifluoromethyl)pyridine-2-carbonitrile
FC(F)(F)C1=CN=C(C=C1)C#N
This item is not returnable. View return policy
95727-86-9
C7H3F3N2
172.11
MFCD01001119
WDSCJULUXJSJOX-UHFFFAOYSA-N
5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
1535372
5-(trifluoromethyl)pyridine-2-carbonitrile
FC(F)(F)C1=CN=C(C=C1)C#N

It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Beilstein 13339826
Solubility Information Slightly soluble in water.
Formula Weight 172.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Cyano-5-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'