missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Cyano-3-methylpyridine, 98%

Brand:  Alfa Aesar™ L19456.14

Additional Details : CAS Number : 20970-75-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11388766

  • 1300.00 NOK / 25g


In stock, ready to ship: 1
Add to basket

Description

Description

Specifications

Chemical Identifiers

20970-75-6
118.139
WBXZCDIZXWDPBL-UHFFFAOYSA-N
819928
CC1=C(N=CC=C1)C#N
C7H6N2
MFCD00190585
2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
3-methylpyridine-2-carbonitrile

Specifications

2-Cyano-3-methylpyridine
Characteristic
84°C to 87°C
20970-75-6
MFCD00190585
2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
CC1=C(N=CC=C1)C#N
118.139
118.14
115°C (239°F)
141°C (38mmHg)
25g
C7H6N2
112001
WBXZCDIZXWDPBL-UHFFFAOYSA-N
3-methylpyridine-2-carbonitrile
819928
98%
Videos
SDS Documents