missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7022872
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
2444-36-2
C8H7ClO2
170.592
MFCD00004317
IUJAAIZKRJJZGQ-UHFFFAOYSA-N
2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
17124
2-(2-chlorophenyl)acetic acid
C1=CC=C(C(=C1)CC(=O)O)Cl
This item is not returnable. View return policy
2444-36-2
C8H7ClO2
170.592
MFCD00004317
IUJAAIZKRJJZGQ-UHFFFAOYSA-N
2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
17124
2-(2-chlorophenyl)acetic acid
C1=CC=C(C(=C1)CC(=O)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 94°C to 97°C
Boiling Point 145°C (5 mmHg)
Quantity 10 g
Odor Odorless
Beilstein 1101523
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 2-Chlorophenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'