missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7025460
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
57729-79-0
C7H4ClF3N2O2
240.566
MFCD00042153
JLWRJMVXRUKFPA-UHFFFAOYSA-N
2-chloro-6-nitro-4-trifluoromethyl aniline, 4-amino-3-chloro-5-nitrobenzotrifluoride, 6-chloro-2-nitro-4-trifluoromethyl phenylamine, pubchem2811, 2-chloro-4-trifluoromethyl-6-nitroaniline, 4-amino-3-chloro-5-nitro-benzotrifluoride, 2-chloranyl-6-nitro-4-trifluoromethyl aniline, 2-chloro-6-nitro-4-trifluoromethyl-phenylamine, 2-chloro-6-nitro-4-trifluoromethyl phenylamine
2734081
2-chloro-6-nitro-4-(trifluoromethyl)aniline
C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
This item is not returnable. View return policy
57729-79-0
C7H4ClF3N2O2
240.566
MFCD00042153
JLWRJMVXRUKFPA-UHFFFAOYSA-N
2-chloro-6-nitro-4-trifluoromethyl aniline, 4-amino-3-chloro-5-nitrobenzotrifluoride, 6-chloro-2-nitro-4-trifluoromethyl phenylamine, pubchem2811, 2-chloro-4-trifluoromethyl-6-nitroaniline, 4-amino-3-chloro-5-nitro-benzotrifluoride, 2-chloranyl-6-nitro-4-trifluoromethyl aniline, 2-chloro-6-nitro-4-trifluoromethyl-phenylamine, 2-chloro-6-nitro-4-trifluoromethyl phenylamine
2734081
2-chloro-6-nitro-4-(trifluoromethyl)aniline
C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 63°C to 67°C
Quantity 1 g
UN Number UN2811
Formula Weight 240.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-nitro-4-(trifluoromethyl)aniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'