Learn More
2-chloro-5-isocyanatopyridine, 97%, Thermo Scientific™
Catalog Number p-4510282
Click to view available options
Quantity:
250 mg
1 g
5 g
125117-96-6
C6H3ClN2O
154.55
MFCD07368563
WHENXACISRCOHK-UHFFFAOYSA-N
4961273
2-chloro-5-isocyanatopyridine
ClC1=NC=C(C=C1)N=C=O
125117-96-6
C6H3ClN2O
154.55
MFCD07368563
WHENXACISRCOHK-UHFFFAOYSA-N
4961273
2-chloro-5-isocyanatopyridine
ClC1=NC=C(C=C1)N=C=O
Assay Percent Range 97%
Quantity 250 mg
Synonym pyridine,2-chloro-5-isocyanato, acmc-20anz3, 6-chloropyridin-3-isocyanate, 2-chloro-5-isocyanato-pyridine, 6-chloro-3-pyridinyl isocyanate, pyridine, 2-chloro-5-isocyanato, 2-chloropyridin-5-yl isocyanate, 2-chloro-5-isocyanatopyridine
Formula Weight 154.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-isocyanatopyridine