Learn More
2-Chloro-4-nitrobenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 182.57 |
Percent Purity | 97% |
Chemical Name or Material | 2-chloro-4-nitrobenzonitrile |