missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-methoxyphenylacetonitrile, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents and acids.
Specifications
Melting Point | 41°C to 43°C |
Quantity | 1 g |
UN Number | UN3439 |
Solubility Information | Insoluble in water. |
Formula Weight | 181.62 |
Percent Purity | 95% |
Chemical Name or Material | 2-Chloro-4-methoxyphenylacetonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'