Learn More
Alfa Aesar™ 2-Chloro-4-cyanopyridine, 97%
Brand: Alfa Aesar™ L19858.14
Additional Details : CAS Number : 33252-30-1 Weight : 0.02500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
33252-30-1 | |
138.554 | |
QRXBTPFMCTXCRD-UHFFFAOYSA-N | |
568638 | |
C1=CN=C(C=C1C#N)Cl |
C6H3ClN2 | |
MFCD00174318 | |
2-chloro-4-cyanopyridine, 2-chloroisonicotinonitrile, 2-chloro-4-pyridinecarbonitrile, 2-chloro-isonicotinonitrile, 4-cyano-2-chloropyridine, 4-pyridinecarbonitrile, 2-chloro, 2-chloro-4-cyano pyridine, 2-chlorisonicotinonitril, pubchem2582 | |
2-chloropyridine-4-carbonitrile |
Specifications
2-Chloro-4-cyanopyridine | |
69°C to 72°C | |
33252-30-1 | |
MFCD00174318 | |
113927 | |
QRXBTPFMCTXCRD-UHFFFAOYSA-N | |
2-chloropyridine-4-carbonitrile | |
568638 | |
97% |
104°C to 106°C (15mmHg) | |
25g | |
C6H3ClN2 | |
UN3439 | |
2-chloro-4-cyanopyridine, 2-chloroisonicotinonitrile, 2-chloro-4-pyridinecarbonitrile, 2-chloro-isonicotinonitrile, 4-cyano-2-chloropyridine, 4-pyridinecarbonitrile, 2-chloro, 2-chloro-4-cyano pyridine, 2-chlorisonicotinonitril, pubchem2582 | |
C1=CN=C(C=C1C#N)Cl | |
138.554 | |
138.56 |