Learn More
2-Chloro-3-nitro-4-picoline, 98%, Thermo Scientific™
Catalog Number p-3211635
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
23056-39-5
C6H5ClN2O2
172.57
MFCD00012347
JHARVUVBTAAPLA-UHFFFAOYSA-N
2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
345363
2-chloro-4-methyl-3-nitropyridine
CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
23056-39-5
C6H5ClN2O2
172.57
MFCD00012347
JHARVUVBTAAPLA-UHFFFAOYSA-N
2-chloro-3-nitro-4-picoline, pyridine, 2-chloro-4-methyl-3-nitro, 2-chloro-3-nitro-4-methylpyridine, 2-chloro-4-methyl-3-nitro-pyridine, pubchem6148, acmc-1cui4, 2-chloro-3-nitro4-picoline, ksc496c3n, 2-chlor-4-methyl-3-nitropyridin
345363
2-chloro-4-methyl-3-nitropyridine
CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 53°C
Color Yellow to Beige
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 172.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-3-nitro-4-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3-nitro-4-picoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.