missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylbenzotrifluoride, 95%, Thermo Scientific™

Catalog Number p-4812769
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
74483-48-0
C8H6ClF3
194.58
NJLAMXMEANIPBX-UHFFFAOYSA-N
2-chloro-3-methylbenzotrifluoride, 2-chloro-1-methyl-3-trifluoromethyl benzene, chloro-3-methyl benzotrifluoride
12712336
2-chloro-1-methyl-3-(trifluoromethyl)benzene
CC1=C(C(=CC=C1)C(F)(F)F)Cl
This item is not returnable. View return policy
74483-48-0
C8H6ClF3
194.58
NJLAMXMEANIPBX-UHFFFAOYSA-N
2-chloro-3-methylbenzotrifluoride, 2-chloro-1-methyl-3-trifluoromethyl benzene, chloro-3-methyl benzotrifluoride
12712336
2-chloro-1-methyl-3-(trifluoromethyl)benzene
CC1=C(C(=CC=C1)C(F)(F)F)Cl
Boiling Point 82°C to 84°C (40.0 mBar)
Flash Point 64°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 194.58
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-methylbenzotrifluoride
compliance-icons
Product Identifier
  • 2-Chloro-3-methylbenzotrifluoride, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'