Learn More
2-Butene-d8 (gas) (cis/trans mixture), CDN
Click to view available options
Quantity:
0.25 L
Unit Size:
0.25 L
Specifications
Specifications
| Chemical Name or Material | 2-Butene-D8 |
| CAS | 6157-20-6 |
| Molecular Formula | C4 D8 |
| Quantity | 0.25 L |
| Synonym | 2-Butene-1,1,1,2,3,4,4,4-d8 (8 CI, 9 CI), 2-Butene-d8 (6 CI, 7 CI), 2-Butene-d8 (cis/trans mixture), (EZ)-2-Butene-1,1,1,2,3,4,4,4-d8, 2-Butene-D8 |
| InChI Formula | InChI=1 S/C4H8/c1-3-4-2/h3-4 H,1-2H3/i1D3,2D3,3 D,4 D |
| SMILES | [2 H]C(=C([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,4,4,4-octadeuteriobut-2-ene |
| Molecular Weight (g/mol) | 64.1556 |
| Formula Weight | 64.1128 g/mol |
| Show More |