missing translation for 'onlineSavingsMsg'
Learn More

2-Bromomalonaldehyde, 97+%, Thermo Scientific Chemicals

Catalog Number p-4812758
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2065-75-0
C3H3BrO2
150.95
SURMYNZXHKLDFO-UHFFFAOYSA-N
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
74945
2-bromopropanedial
C(=O)C(C=O)Br
This item is not returnable. View return policy
2065-75-0
C3H3BrO2
150.95
SURMYNZXHKLDFO-UHFFFAOYSA-N
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
74945
2-bromopropanedial
C(=O)C(C=O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 132.0°C to 136.0°C
Assay Percent Range 98.5 % min. (Acid)
Packaging Glass bottle
Linear Formula BrCH(CHO)2
Quantity 1 g
Formula Weight 150.95
Percent Purity 97+%
Chemical Name or Material 2-Bromomalonaldehyde
compliance-icons compliance-icons
Product Identifier
  • 2-Bromomalonaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'