missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromobenzylamine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Pharmaceutical intermediates. As intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pharmaceutical intermediates. As intermediate in organic synthesis.
Solubility
Insoluble in Water. Soluble in Alcohol and Ether.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..
Pharmaceutical intermediates. As intermediate in organic synthesis.
Solubility
Insoluble in Water. Soluble in Alcohol and Ether.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide..
Specifications
Boiling Point | 118°C to 119°C (9 mmHg) |
Odor | Amine-like |
Quantity | 5 g |
UN Number | UN2735 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in Water. Soluble in Alcohol and Ether. |
Formula Weight | 186.06 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromobenzylamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'