missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-N-methylaniline, 95%, Thermo Scientific Chemicals

Catalog Number 15431328
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6832-87-7
C7H8BrN
186.052
MFCD06798064
SMVIAQFTVWDWDS-UHFFFAOYSA-N
334086
2-bromo-N-methylaniline
CNC1=CC=CC=C1Br
This item is not returnable. View return policy
6832-87-7
C7H8BrN
186.052
MFCD06798064
SMVIAQFTVWDWDS-UHFFFAOYSA-N
334086
2-bromo-N-methylaniline
CNC1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Density 1.589 g/mL
Boiling Point 107°C to 109°C (12 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.607
Quantity 250 mg
Synonym 2-bromo-n-methylbenzenamine, acmc-20abqa, o-bromo-n-methylaniline, n-methyl-2-bromoaniline, benzenamine,2-bromo-n-methyl, 2-bromo-n-methylaniline, benzenamine, 2-bromo-n-methyl
Formula Weight 186.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-N-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'