Learn More
2-Bromo-6-fluoroaniline, 95%, Thermo Scientific™
Catalog Number p-4090018
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
65896-11-9
C6H5BrFN
190.02
MFCD01310982
ALZFPYUPNVLVQM-UHFFFAOYSA-N
benzenamine, 2-bromo-6-fluoro, 2-bromo-6-fluoro-phenylamine, 2-bromo-6-fluorobenzenamine, 2-bromo-6-fluoro-aniline, 2-bromo-6-fluoro aniline, 2-bromo-6-fluorophenylamine, pubchem2923, acmc-209nsu, 2-bromo-6-flouroaniline, ksc493o8h
2782940
2-bromo-6-fluoroaniline
NC1=C(F)C=CC=C1Br
65896-11-9
C6H5BrFN
190.02
MFCD01310982
ALZFPYUPNVLVQM-UHFFFAOYSA-N
benzenamine, 2-bromo-6-fluoro, 2-bromo-6-fluoro-phenylamine, 2-bromo-6-fluorobenzenamine, 2-bromo-6-fluoro-aniline, 2-bromo-6-fluoro aniline, 2-bromo-6-fluorophenylamine, pubchem2923, acmc-209nsu, 2-bromo-6-flouroaniline, ksc493o8h
2782940
2-bromo-6-fluoroaniline
NC1=C(F)C=CC=C1Br
CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.6740g/mL
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(F)NH2
Refractive Index 1.5810 to 1.5830 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.674
Formula Weight 190.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-fluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-6-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.