Learn More
2-Bromo-5-nitrothiazole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Bromo-5-nitrothiazole is used as a building block. It is used as an intermediate to produce other chemicals.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 85°C to 90°C |
Quantity | 1 g |
Beilstein | 4856 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 209.03 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-5-nitrothiazole |