missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylbenzoic acid, 98%, Thermo Scientific™

Catalog Number p-7077720
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7697-27-0
C8H7BrO2
215.05
MFCD00045797
ZZYYOHPHSYCHQG-UHFFFAOYSA-N
4-methyl-2-bromobenzoic acid, 2-bromo-p-toluic acid, 2-bromo-4-methyl-benzoic acid, benzoic acid, 2-bromo-4-methyl, 2-bromo-4-methyl benzoic acid, zlchem 427, pubchem4719, 3-bromo-4-carboxytoluene, acmc-1bbb4, 2-bromo-4-methylbenzoicacid
226292
2-bromo-4-methylbenzoic acid
CC1=CC=C(C(O)=O)C(Br)=C1
This item is not returnable. View return policy
7697-27-0
C8H7BrO2
215.05
MFCD00045797
ZZYYOHPHSYCHQG-UHFFFAOYSA-N
4-methyl-2-bromobenzoic acid, 2-bromo-p-toluic acid, 2-bromo-4-methyl-benzoic acid, benzoic acid, 2-bromo-4-methyl, 2-bromo-4-methyl benzoic acid, zlchem 427, pubchem4719, 3-bromo-4-carboxytoluene, acmc-1bbb4, 2-bromo-4-methylbenzoicacid
226292
2-bromo-4-methylbenzoic acid
CC1=CC=C(C(O)=O)C(Br)=C1
Melting Point 144.0°C to 145.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-methylbenzoic acid
compliance-icons
Product Identifier
  • 2-Bromo-4-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'