missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroaniline, 98%, Thermo Scientific™

Catalog Number p-3593241
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
873-38-1
C6H5BrClN
206.47
MFCD00041313
SYTBIFURTZACKR-UHFFFAOYSA-N
benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline
70110
2-bromo-4-chloroaniline
C1=CC(=C(C=C1Cl)Br)N
This item is not returnable. View return policy
873-38-1
C6H5BrClN
206.47
MFCD00041313
SYTBIFURTZACKR-UHFFFAOYSA-N
benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline
70110
2-bromo-4-chloroaniline
C1=CC(=C(C=C1Cl)Br)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 64°C to 68°C
Color Brown
Boiling Point 127°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(Br)NH2
Quantity 1 g
Formula Weight 206.47
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-chloroaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-4-chloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'