missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-hexylthiophene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agentsstrong acids and bases.
2-Bromo-3-hexylthiophene is used in the synthesis of End-capped Regioregular Poly(3-hexylthiophene). It is also used to synthesize well-defined head-to-tail-type oligothiophenes which are used in a number of high-technology applications including OLEDs.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agentsstrong acids and bases.
Specifications
Density | 1.24 |
Flash Point | 110°C (230°F) |
Refractive Index | 1.529 |
Quantity | 5 g |
Solubility Information | Slightly miscible with water. |
Formula Weight | 247.2 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-3-hexylthiophene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'