missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(2-thienyl)-1-ethanone, Technical Grade, Thermo Scientific™

Catalog Number p-4510096
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10531-41-6
C6H5BrOS
205.069
UHWNENCHFSDZQP-UHFFFAOYSA-N
2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
2776372
2-bromo-1-thiophen-2-ylethanone
C1=CSC(=C1)C(=O)CBr
This item is not returnable. View return policy
10531-41-6
C6H5BrOS
205.069
UHWNENCHFSDZQP-UHFFFAOYSA-N
2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
2776372
2-bromo-1-thiophen-2-ylethanone
C1=CSC(=C1)C(=O)CBr
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.07
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(2-thienyl)-1-ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'