Learn More
2-Aminopyrazine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
50 g
Description
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 118°C to 121°C |
Quantity | 5 g |
Beilstein | 107025 |
Solubility Information | Soluble in water. |
Formula Weight | 95.11 |
Percent Purity | 99% |
Chemical Name or Material | 2-Aminopyrazine |