Learn More
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%, Thermo Scientific Chemicals
Catalog Number p-7046259
Click to view available options
Quantity:
100 mg
1 g
80983-34-2
C10H13N3O2S
239.29
MFCD01075656
WTPBIYSMFKUQKY-UHFFFAOYSA-N
albendazole-2-aminosulfone, 2-amino-5-propylsulphonylbenzimidazole, 2-amino-5-n-propylsulfonylbenzimidazole, 1h-benzimidazol-2-amine, 5-propylsulfonyl, 5-propylsulfonyl-1h-benzo d imidazol-2-amine, 2-amino-5-n-propylsulphonylbenzimidazole, abz2nh2, 5-propylsulfonyl-1h-benzimidazol-2-amine, 5-propylsulphonyl-1h-benzimidazol-2-amine, 5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-amine
88125
CHEBI:80621
6-propylsulfonyl-1H-benzimidazol-2-amine
CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1
80983-34-2
C10H13N3O2S
239.29
MFCD01075656
WTPBIYSMFKUQKY-UHFFFAOYSA-N
albendazole-2-aminosulfone, 2-amino-5-propylsulphonylbenzimidazole, 2-amino-5-n-propylsulfonylbenzimidazole, 1h-benzimidazol-2-amine, 5-propylsulfonyl, 5-propylsulfonyl-1h-benzo d imidazol-2-amine, 2-amino-5-n-propylsulphonylbenzimidazole, abz2nh2, 5-propylsulfonyl-1h-benzimidazol-2-amine, 5-propylsulphonyl-1h-benzimidazol-2-amine, 5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-amine
88125
CHEBI:80621
6-propylsulfonyl-1H-benzimidazol-2-amine
CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 220°C to 222°C
Quantity 100 mg
Formula Weight 239.3
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-n-propylsulfonylbenzimidazole