missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3,4-dimethylpyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7047126
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
374537-97-0
C7H9BrN2
201.07
MFCD02093947
YAVKJNIMFGZBSY-UHFFFAOYSA-N
2-amino-5-bromo-3,4-dimethylpyridine, 5-bromo-3,4-dimethyl-2-pyridinamine, acmc-20agep, 5-bromo-3,4-dimethyl-2-pyridylamine, 2-pyridinamine,5-bromo-3,4-dimethyl, 5-bromo-3,4-dimethyl-pyridin-2-ylamine
7010307
5-bromo-3,4-dimethylpyridin-2-amine
CC1=C(C)C(N)=NC=C1Br
This item is not returnable. View return policy
374537-97-0
C7H9BrN2
201.07
MFCD02093947
YAVKJNIMFGZBSY-UHFFFAOYSA-N
2-amino-5-bromo-3,4-dimethylpyridine, 5-bromo-3,4-dimethyl-2-pyridinamine, acmc-20agep, 5-bromo-3,4-dimethyl-2-pyridylamine, 2-pyridinamine,5-bromo-3,4-dimethyl, 5-bromo-3,4-dimethyl-pyridin-2-ylamine
7010307
5-bromo-3,4-dimethylpyridin-2-amine
CC1=C(C)C(N)=NC=C1Br

It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 133°C to 135°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 201.07
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3,4-dimethylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'