Learn More
2-Amino-4-cyanopyridine, 97%, Thermo Scientific Chemicals
Catalog Number p-7047848
Click to view available options
Quantity:
1 g
5 g
25 g
42182-27-4
C6H5N3
119.127
MFCD03791310
GEEAYLFEIFJFGP-UHFFFAOYSA-N
2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine
7015524
2-aminopyridine-4-carbonitrile
C1=CN=C(C=C1C#N)N
42182-27-4
C6H5N3
119.127
MFCD03791310
GEEAYLFEIFJFGP-UHFFFAOYSA-N
2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine
7015524
2-aminopyridine-4-carbonitrile
C1=CN=C(C=C1C#N)N
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 148°C to 152°C
Quantity 1 g
UN Number UN3439
Beilstein 386393
Solubility Information Slightly soluble in water.
Formula Weight 119.13
Percent Purity 97%
Chemical Name or Material 2-Amino-4-cyanopyridine