Learn More
Alfa Aesar™ 2-Amino-4-cyanopyridine, 97%
Brand: Alfa Aesar™ L19841.14
Additional Details : CAS Number : 42182-27-4 Weight : 0.02500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 25g |
---|
Description
Applications2-Amino-4-cyanopyridine is used in the treatment of central nervous system (CNS) disorders and as a mGluR2 antagonist.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
42182-27-4 | |
119.127 | |
GEEAYLFEIFJFGP-UHFFFAOYSA-N | |
7015524 | |
C1=CN=C(C=C1C#N)N |
C6H5N3 | |
MFCD03791310 | |
2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine | |
2-aminopyridine-4-carbonitrile |
Specifications
2-Amino-4-cyanopyridine | |
25g | |
C6H5N3 | |
UN3439 | |
2-amino-4-cyanopyridine, 2-aminoisonicotinonitrile, 2-amino-isonicotinonitrile, pubchem7664, 4-cyanopyridin-2-amine, 4-cyano-2-aminopyridine, acmc-1art6, 2-amino-4-cyano-pyridine, ksc490s3f, 4-cyano-2-pyridinamine | |
GEEAYLFEIFJFGP-UHFFFAOYSA-N | |
2-aminopyridine-4-carbonitrile | |
7015524 | |
97% |
148°C to 152°C | |
42182-27-4 | |
MFCD03791310 | |
386393 | |
Slightly soluble in water. | |
C1=CN=C(C=C1C#N)N | |
119.127 | |
119.13 |